H-Cys(Z)-OH 95%

Supplier: Ambeed
Warning

Synonyms: H-Cys(Cbz)-OH

A214549-100G A214549-1G A214549-25G A214549-10G A214549-5G
76999-718EA 215 USD
76999-718 76999-720 76999-722 77881-522 77881-523
H-Cys(Z)-OH 95%
H-Cys(Z)-OH

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₄S
MW: 255.3 g/mol
MDL Number: MFCD00055774
CAS Number: 1625-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR