7-Bromo-1-methyl-1,3-dihydro-2H-benzo[d]imidazol-2-one 95%

Supplier: Ambeed
Warning

A214455-100MG A214455-1G A214455-250MG A214455-50MG A214455-5G
77013-038EA 112.46 USD
77013-038 77013-040 77013-042 77013-044 77013-046
7-Bromo-1-methyl-1,3-dihydro-2H-benzo[d]imidazol-2-one 95%
7-Bromo-1-methyl-1,3-dihydro-2H-benzo[d]imidazol-2-one

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O
MW: 227.05 g/mol
MDL Number: MFCD27932420
CAS Number: 913297-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR