4-Amino-6-chloropyrimidin-5-ol 96%

Supplier: Ambeed
Warning

A214447-100MG A214447-1G A214447-250MG A214447-5G
77030-600EA 26 USD
77030-600 77030-602 77030-604 77872-095
4-Amino-6-chloropyrimidin-5-ol 96%
4-Amino-6-chloropyrimidin-5-ol

Precaución: for Research Use Only

Formula: C₄H₄ClN₃O
MDL Number: MFCD13689013
CAS Number: 38953-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR