5-Bromo-3-phenyl-1H-indazole 95%

Supplier: Ambeed
Warning

A214442-100MG A214442-250MG
76986-860EA 100.79 USD
76986-860 76986-862
5-Bromo-3-phenyl-1H-indazole 95%
5-Bromo-3-phenyl-1H-indazole

Precaución: for Research Use Only

Formula: C₁₃H₉BrN₂
MW: 273.13 g/mol
MDL Number: MFCD08549635
CAS Number: 57639-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR