(3-Chloro-4-methoxyphenyl)methanamine 98%

Supplier: Ambeed
Danger

Synonyms: 3-Chloro-4-methoxybenzylamine

A214158-25G A214158-5G
76999-710EA 65.78 USD
76999-710 76999-712
(3-Chloro-4-methoxyphenyl)methanamine 98%
(3-Chloro-4-methoxyphenyl)methanamine

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO
MW: 171.63 g/mol
MDL Number: MFCD00995395
CAS Number: 115514-77-7
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR