Dodecyl 4,6-dibromothieno[3,4-b]thiophene-2-carboxylate 95%

Supplier: Ambeed
Warning

A214135-100MG A214135-1G A214135-250MG
76986-842EA 9.55 USD
76986-842 76986-844 76986-846
Dodecyl 4,6-dibromothieno[3,4-b]thiophene-2-carboxylate 95%
Dodecyl 4,6-dibromothieno[3,4-b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₉H₂₆Br₂O₂S₂
MW: 510.35 g/mol
MDL Number: MFCD16619393
CAS Number: 1098102-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR