1-Methyl-1H-indole-4-carbaldehyde 95%

Supplier: Ambeed
Warning

A214069-1G A214069-250MG A214069-5G
77030-594EA 220.67 USD
77030-594 77030-596 77030-598
1-Methyl-1H-indole-4-carbaldehyde 95%
1-Methyl-1H-indole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MDL Number: MFCD08690261
CAS Number: 133994-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR