4-Amino-3,5-dichloropyridazine 98%

Supplier: Ambeed
Warning

A213955-100MG A213955-1G A213955-250MG
77030-588EA 21 USD
77030-588 77030-590 77030-592
4-Amino-3,5-dichloropyridazine 98%
4-Amino-3,5-dichloropyridazine

Precaución: for Research Use Only

Formula: C₄H₃Cl₂N₃
MDL Number: MFCD01647251
CAS Number: 53180-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR