3-Bromo-5-fluoro-4-nitrobenzonitrile 95%

Supplier: Ambeed
Warning

A213714-100MG A213714-250MG A213714-1G
77013-020EA 37 USD
77013-020 77013-022 77593-390
3-Bromo-5-fluoro-4-nitrobenzonitrile 95%
3-Bromo-5-fluoro-4-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂BrFN₂O₂
MDL Number: MFCD26743737
CAS Number: 1310918-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR