Methyl 4-bromo-3,5-dimethylbenzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-3,5-dimethylbenzoic acid methyl ester
A213671-100MG
A213671-1G
A213671-250MG
A213671-5G
76986-816EA
23.34
USD
76986-816
76986-818
76986-820
76986-822
Methyl 4-bromo-3,5-dimethylbenzoate 97%
Methyl 4-bromo-3,5-dimethylbenzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁BrO₂ MW: 243.10 g/mol |
MDL Number:
MFCD09833419 CAS Number: 432022-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...