D-(-)-Arabinose 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
D-(-)-Arabinose, (2S,3R,4R)-2,3,4,5-Tetrahydroxypentanal
A213618-100G
A213618-10G
A213618-500G
A213618-25G
76986-810EA
28.65
USD
76986-810
76986-812
76986-814
77683-608
D-(-)-Arabinose 97%
D(-)-Arabinose
Precaución: For research use only.
Formula:
C₅H₁₀O₅ MW: 150.13 g/mol |
MDL Number:
MFCD00135608 CAS Number: 10323-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...