2-Chloro-6-methoxy-4-methylpyridine 97%

Supplier: Ambeed
Warning

A213553-1G A213553-250MG A213553-5G
77013-016EA 53 USD
77013-016 77890-271 77890-272
2-Chloro-6-methoxy-4-methylpyridine 97%
2-Chloro-6-methoxy-4-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₈ClNO
MDL Number: MFCD16610595
CAS Number: 25297-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR