5-Methoxy-1H-indole-2-carbaldehyde 95%

Supplier: Ambeed
Warning

A213514-1G A213514-250MG
76999-676EA 244.01 USD
76999-676 76999-678
5-Methoxy-1H-indole-2-carbaldehyde 95%
5-Methoxy-1H-indole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.18 g/mol
MDL Number: MFCD00297147
CAS Number: 21778-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR