2-Amino-4-hydroxy-6-(trifluoromethyl)pyrimidine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-hydroxy-6-trifluoromethylpyrimidine

A213477-10G A213477-1G A213477-25G A213477-5G A213477-100G
77013-008EA 51 USD
77013-008 77013-010 77013-012 77013-014 77890-268
2-Amino-4-hydroxy-6-(trifluoromethyl)pyrimidine 95%
2-Amino-4-hydroxy-6-(trifluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₄F₃N₃O
MDL Number: MFCD00068093
CAS Number: 1513-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR