4-Chloro-1H-indazol-5-amine 98%

Supplier: Ambeed
Warning

A213447-100MG A213447-250MG
76986-800EA 42.44 USD
76986-800 76986-802
4-Chloro-1H-indazol-5-amine 98%
4-Chloro-1H-indazol-5-amine

Precaución: for Research Use Only

Formula: C₇H₆ClN₃
MW: 167.59 g/mol
MDL Number: MFCD11007840
CAS Number: 1082041-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR