2,5-Dichloro-4-methylpyridine 97%

Supplier: Ambeed
Warning

A213442-10G A213442-1G A213442-25G A213442-5G A213442-100G
77030-568EA 82.76 USD
77030-568 77030-570 77030-572 77030-574 77667-790
2,5-Dichloro-4-methylpyridine 97%
2,5-Dichloro-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅Cl₂N
MW: 162.0166 g/mol
MDL Number: MFCD07375099
CAS Number: 886365-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR