Fmoc-2-chloro-D-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Phe(2-Cl)-OH

A213076-1G A213076-250MG A213076-5G
76999-656EA 27 USD
76999-656 76999-658 77671-898
Fmoc-2-chloro-D-phenylalanine 97%
Fmoc-2-chloro-D-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₀ClNO₄
MW: 421.88 g/mol
MDL Number: MFCD00672547
CAS Number: 205526-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR