2-Bromo-4-methoxy-1-nitrobenzene 98%

Supplier: Ambeed
Warning

A213057-100MG A213057-1G A213057-5G A213057-10G
76986-792EA 7.43 USD
76986-792 76986-794 76986-796 77683-606
2-Bromo-4-methoxy-1-nitrobenzene 98%
2-Bromo-4-methoxy-1-nitrobenzene

Precaución: For research use only.

Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
MDL Number: MFCD06797977
CAS Number: 98447-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR