6-Bromo-2-chloroquinazolin-4-amine 97%

Supplier: Ambeed
Warning

A213044-100MG A213044-1G A213044-5G
77030-562EA 25.47 USD
77030-562 77600-718 77667-784
6-Bromo-2-chloroquinazolin-4-amine 97%
6-Bromo-2-chloroquinazolin-4-amine

Precaución: for Research Use Only

Formula: C₈H₅BrClN₃
MW: 258.50 g/mol
MDL Number: MFCD05739655
CAS Number: 111218-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR