2-Isopropyl-1,3-thiazole-4-carbaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 2-Isopropylthiazole-4-carbaldehyde

A212694-100MG A212694-1G A212694-250MG
77030-552EA 45.62 USD
77030-552 77030-554 77030-556
2-Isopropyl-1,3-thiazole-4-carbaldehyde 95%
2-Isopropyl-1,3-thiazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₉NOS
MW: 155.22 g/mol
MDL Number: MFCD06660125
CAS Number: 133047-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR