N-Cyclopropyl-3-nitropyridin-2-amine 98%

Supplier: Ambeed
Warning

A212687-1G A212687-250MG A212687-5G
76999-650EA 8 USD
76999-650 76999-652 76999-654
N-Cyclopropyl-3-nitropyridin-2-amine 98%
N-Cyclopropyl-3-nitropyridin-2-amine

Precaución: for Research Use Only

Formula: C₈H₉N₃O₂
MDL Number: MFCD00174356
CAS Number: 290313-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR