3-Chloro-6-(piperazin-1-yl)pyridazine hydrochloride 95%

Supplier: Ambeed
Warning

A212352-1G A212352-250MG
77012-984EA 23.34 USD
77012-984 77012-986
3-Chloro-6-(piperazin-1-yl)pyridazine hydrochloride 95%
3-Chloro-6-(piperazin-1-yl)pyridazine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂Cl₂N₄
MW: 235.11 g/mol
MDL Number: MFCD16620387
CAS Number: 100241-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR