2-Piperidin-1-ylmethylbenzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Piperidin-1-ylmethyl)benzoic acid

A212283-100MG A212283-250MG
76999-642EA 78.51 USD
76999-642 76999-644
2-Piperidin-1-ylmethylbenzoic acid 95%
2-Piperidin-1-ylmethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₂
MW: 219.29 g/mol
MDL Number: MFCD08690199
CAS Number: 917747-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR