(3-Chloropyridin-2-yl)methanol 97%

Supplier: Ambeed
Warning

A212200-10G A212200-1G A212200-250MG A212200-5G A212200-100MG
76986-756EA 387 USD
76986-756 76986-758 76986-760 76986-762 77890-266
(3-Chloropyridin-2-yl)methanol 97%
(3-Chloropyridin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₆ClNO
MDL Number: MFCD11110226
CAS Number: 60588-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR