Adamantan-1-yl acrylate 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

A212098-1G A212098-5G A212098-250MG
77030-546EA 45 USD
77030-546 77667-772 77872-082
Adamantan-1-yl acrylate 98% stabilized
Adamantan-1-yl acrylate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₂
MDL Number: MFCD18072700
CAS Number: 121601-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR