2-Bromo-6-fluoro-3-methylphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (2-Bromo-6-fluoro-3-methylphenyl)boronic acid

A211845-1G A211845-250MG A211845-5G
76999-632EA 25.47 USD
76999-632 76999-634 76999-636
2-Bromo-6-fluoro-3-methylphenylboronic acid 97%
2-Bromo-6-fluoro-3-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrFO₂
MW: 232.8400 g/mol
MDL Number: MFCD09800874
CAS Number: 957121-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR