Spiro[indoline-3,4'-piperidin]-2-one hydrochloride 98%

Supplier: Ambeed
Warning

A211800-1G A211800-250MG
77030-530EA 158.08 USD
77030-530 77030-532
Spiro[indoline-3,4'-piperidin]-2-one hydrochloride 98%
Spiro[indoline-3,4'-piperidin]-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClN₂O
MW: 238.71 g/mol
MDL Number: MFCD15071694
CAS Number: 356072-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR