4-Amino-3-nitrobenzophenone 97%

Supplier: Ambeed
Warning

Synonyms: (4-Amino-3-nitrophenyl)(phenyl)methanone

A211738-100G A211738-10G A211738-25G A211738-500G
76999-624EA 65.78 USD
76999-624 76999-626 76999-628 76999-630
4-Amino-3-nitrobenzophenone 97%
4-Amino-3-nitrobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₁₀N₂O₃
MW: 242.23 g/mol
MDL Number: MFCD00007154
CAS Number: 31431-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR