(R)-1-(Pyridin-3-yl)ethanamine 97%

Supplier: Ambeed
Warning

A211732-100MG A211732-250MG A211732-50MG A211732-5G A211732-1G
77012-974EA 55.17 USD
77012-974 77012-976 77012-978 77012-980 77683-603
(R)-1-(Pyridin-3-yl)ethanamine 97%
(R)-1-(Pyridin-3-yl)ethanamine

Precaución: For research use only.

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD09256802
CAS Number: 40154-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR