tert-Butyl 3-(3-methylpyridin-2-yl)benzoate 97%
Supplier: Ambeed
|
Warning
|
A211665-100MG
A211665-1G
A211665-250MG
76986-750EA
36.08
USD
76986-750
76986-752
76986-754
tert-Butyl 3-(3-methylpyridin-2-yl)benzoate 97%
tert-Butyl 3-(3-methylpyridin-2-yl)benzoate
Precaución: for Research Use Only
Formula:
C₁₇H₁₉NO₂ MW: 269.34 g/mol |
MDL Number:
MFCD26383927 CAS Number: 1083057-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...