6-Chloro-2-methylpyrimidin-4-amine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-6-chloro-2-methylpyrimidine, 4-Pyrimidinamine, 6-chloro-2-methyl-, 6-chloro-2-methylpyrimidin-4-amine

A211484-100G A211484-10G A211484-1G A211484-25G A211484-500G A211484-5G
76986-734EA 279.02 USD
76986-734 76986-736 76986-738 76986-740 76986-742 76986-744
6-Chloro-2-methylpyrimidin-4-amine 97%
6-Chloro-2-methylpyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.58 g/mol
MDL Number: MFCD07782029
CAS Number: 1749-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR