Ethyl 1H-pyrrole-3-carboxylate 98%

Supplier: Ambeed
Warning

A211273-1G A211273-250MG A211273-5G
77030-500EA 108.22 USD
77030-500 77030-502 77030-504
Ethyl 1H-pyrrole-3-carboxylate 98%
Ethyl 1H-pyrrole-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MW: 139.15 g/mol
MDL Number: MFCD10000696
CAS Number: 37964-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR