3,5-Dimethyl-4-nitrophenol 97%

Supplier: Ambeed
Warning

A211216-10G A211216-1G A211216-250MG A211216-25G A211216-5G
76986-724EA 22 USD
76986-724 76986-726 76986-728 76986-730 76986-732
3,5-Dimethyl-4-nitrophenol 97%
3,5-Dimethyl-4-nitrophenol

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MDL Number: MFCD00031365
CAS Number: 5344-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR