2-Fluoro-3-iodobenzaldehyde 95%

Supplier: Ambeed
Warning

A211178-100MG A211178-1G A211178-250MG A211178-5G
77012-964EA 26.53 USD
77012-964 77012-966 77012-968 77012-970
2-Fluoro-3-iodobenzaldehyde 95%
2-Fluoro-3-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄FIO
MW: 250.009 g/mol
MDL Number: MFCD15528731
CAS Number: 146137-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR