(S)-1-Boc-Piperazine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid, (2S)-1-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid

A210960-1G A210960-250MG A210960-25G A210960-5G
76999-610EA 98.67 USD
76999-610 76999-612 76999-614 76999-616
(S)-1-Boc-Piperazine-2-carboxylic acid 97%
(S)-1-Boc-Piperazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₄
MW: 230.2600 g/mol
MDL Number: MFCD02179105
CAS Number: 159532-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR