6-Hydroxypyridazine-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A210954-100G A210954-10G A210954-1G A210954-250MG A210954-5G A210954-25G
77012-950EA 360.71 USD
77012-950 77012-952 77012-954 77012-956 77012-958 77683-601
6-Hydroxypyridazine-3-carboxylic acid 98%
6-Hydroxypyridazine-3-carboxylic acid

Precaución: For research use only.

Formula: C₅H₄N₂O₃
MW: 140.1000 g/mol
MDL Number: MFCD00052805
CAS Number: 37972-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR