1H-Indazole-6-carbonitrile 98%

Supplier: Ambeed
Warning

A210599-100MG A210599-250MG A210599-5G A210599-1G
77012-940EA 10 USD
77012-940 77012-942 77012-944 77890-256
1H-Indazole-6-carbonitrile 98%
1H-Indazole-6-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₅N₃
MDL Number: MFCD04973398
CAS Number: 141290-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR