1-Methyl-1H-indole-6-boronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole, 2-(1-Methyl-1H-indol-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A210399-100MG A210399-1G A210399-250MG A210399-10G A210399-5G
77030-480EA 21 USD
77030-480 77030-482 77030-484 77872-069 77872-070
1-Methyl-1H-indole-6-boronic acid, pinacol ester 98%
1-Methyl-1H-indole-6-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₀BNO₂
MW: 257.136 g/mol
MDL Number: MFCD08690255
CAS Number: 884507-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR