1-(2-Fluoro-6-(trifluoromethyl)benzyl)-6-methylpyrimidine-2,4(1H,3H)-dione 95%

Supplier: Ambeed
Warning

A210259-100MG A210259-1G A210259-250MG A210259-5G
77012-922EA 6.37 USD
77012-922 77012-924 77012-926 77012-928
1-(2-Fluoro-6-(trifluoromethyl)benzyl)-6-methylpyrimidine-2,4(1H,3H)-dione 95%
1-(2-Fluoro-6-(trifluoromethyl)benzyl)-6-methylpyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₄N₂O₂
MW: 302.22 g/mol
MDL Number: MFCD12546645
CAS Number: 830346-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR