4-Bromo-1,1':3',1''-terphenyl 98%

Supplier: Ambeed
Warning

A210243-1G A210243-250MG A210243-5G
77012-916EA 18 USD
77012-916 77012-918 77012-920
4-Bromo-1,1':3',1''-terphenyl 98%
4-Bromo-1,1':3',1''-terphenyl

Precaución: for Research Use Only

Formula: C₁₈H₁₃Br
MDL Number: MFCD20486761
CAS Number: 54590-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR