5-Amino-6-chloropyridine-3-boronic acid, pinacol ester 99%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine, 3-Amino-2-chloropyridine-5-boronic acid, pinacol ester, 3-Amino-2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A210192-1G A210192-250MG
76986-706EA 58 USD
76986-706 76986-708
5-Amino-6-chloropyridine-3-boronic acid, pinacol ester 99%
5-Amino-6-chloropyridine-3-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₆BClN₂O₂
MDL Number: MFCD09260487
CAS Number: 1073354-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR