(R)-2-(Aminomethyl)-3-methylbutanoic acid 97%

Supplier: Ambeed
Warning

A210116-100MG A210116-1G A210116-250MG A210116-5G
76986-700EA 29 USD
76986-700 76986-702 76986-704 77885-515
(R)-2-(Aminomethyl)-3-methylbutanoic acid 97%
(R)-2-(Aminomethyl)-3-methylbutanoic acid

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MDL Number: MFCD07372878
CAS Number: 210345-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR