1-Bromo-2-chloro-4,5-difluorobenzene 98%

Supplier: Ambeed
Warning

A210031-250MG A210031-5G A210031-1G
76986-696EA 33 USD
76986-696 76986-698 77593-368
1-Bromo-2-chloro-4,5-difluorobenzene 98%
1-Bromo-2-chloro-4,5-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClF₂
MDL Number: MFCD00142586
CAS Number: 59447-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR