Ethyl cyclopentylideneacetate 95+%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-cyclopentylideneacetate

A209951-100MG A209951-1G A209951-250MG
77030-470EA 43 USD
77030-470 77030-472 77030-474
Ethyl cyclopentylideneacetate 95+%
Ethyl cyclopentylideneacetate

Precaución: for Research Use Only

Formula: C₉H₁₄O₂
MW: 154.21 g/mol
MDL Number: MFCD11976268
CAS Number: 1903-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR