(2-Ethyl-1H-imidazol-1-yl)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Ethyl-1H-imidazol-1-yl)acetic acid

A209775-100MG A209775-250MG
77030-466EA 26.53 USD
77030-466 77030-468
(2-Ethyl-1H-imidazol-1-yl)acetic acid 95%
(2-Ethyl-1H-imidazol-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.167 g/mol
MDL Number: MFCD07186426
CAS Number: 883539-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR