1-(5-Methoxypyridin-3-yl)ethanone 95%

Supplier: Ambeed
Warning

A209334-100MG A209334-250MG
76986-674EA 56.23 USD
76986-674 76986-676
1-(5-Methoxypyridin-3-yl)ethanone 95%
1-(5-Methoxypyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MDL Number: MFCD07375082
CAS Number: 886364-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR