Benzyl 4-((methylsulfonyl)oxy)piperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A209243-100G A209243-1G A209243-25G A209243-5G
76986-658EA 504.99 USD
76986-658 76986-660 76986-662 76986-664
Benzyl 4-((methylsulfonyl)oxy)piperidine-1-carboxylate 98%
Benzyl 4-((methylsulfonyl)oxy)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₅S
MW: 313.36 g/mol
MDL Number: MFCD09953312
CAS Number: 199103-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR