1-Bromo-4-(2,2,2-trifluoroethyl)benzene 95%

Supplier: Ambeed
Warning

A209218-250MG A209218-25G A209218-5G A209218-1G
76999-570EA 41 USD
76999-570 76999-572 76999-574 77593-360
1-Bromo-4-(2,2,2-trifluoroethyl)benzene 95%
1-Bromo-4-(2,2,2-trifluoroethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₆BrF₃
MDL Number: MFCD10697865
CAS Number: 155820-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR