1-(6-Fluoropyridin-3-yl)ethanone 95%

Supplier: Ambeed
Warning

A209102-1G A209102-250MG A209102-5G
76986-646EA 30 USD
76986-646 76986-648 76986-650
1-(6-Fluoropyridin-3-yl)ethanone 95%
1-(6-Fluoropyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₆FNO
MDL Number: MFCD07375076
CAS Number: 84331-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR