4-Bromo-2,3-difluorophenol 98%

Supplier: Ambeed
Warning

A209076-10G A209076-1G A209076-25G A209076-5G
76999-562EA 40.32 USD
76999-562 76999-564 76999-566 76999-568
4-Bromo-2,3-difluorophenol 98%
4-Bromo-2,3-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrF₂O
MW: 208.99 g/mol
MDL Number: MFCD08061907
CAS Number: 144292-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR